missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Aquacide II

Catalog No. 178515KG
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
178515KG 5 kg
178511KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 178515KG Supplier MilliporeSigma™ Supplier No. 178515KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly viscous salt useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing

  • Sodium salt of carboxymethylcellulose
  • High viscosity
  • Useful for concentration of proteins and other macromolecules and can be used for rapid removal of water from dialysis tubing by using either a 2% syrup or the Aquacide solid
  • Neither denature proteins nor change the pH during concentration
  • At room temperature 1g of Aquacide absorbs about 5mL of water from solution in 1h

Specifications

CAS 9004-32-4
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 500000g/mol
Quantity 5 kg
Form Solid