missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethyl Cellulose Sodium Salt (n=approx. 1,050), TCI America™

Catalog No. C060325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C060325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C060325G Supplier TCI America Supplier No. C060325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use as laboratory reagent.

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Quantity 25 g
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Carboxymethyl Cellulose Sodium Salt (n=approx. 1,050)
EINECSNumber (8)-0203
RTECSNumber FJ5950000
TSCA Yes