missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethyl Cellulose, Sodium Salt, MP Biomedicals™

Catalog No. MP215056192
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215056192 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215056192 Supplier MP Biomedicals Inc Supplier No. 0215056192

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a suspending agent, stabilizer and viscosity-increasing agent. Carboxymethyl Cellulose, Sodium Salt, MP Biomedicals is a high viscosity carboxymethylcellulose (CMC). The viscosity is both concentration and temperature dependent.

The viscosity of is CMC is both concentration and temperature dependent. The viscosity of a 1% solution in water at 25°C is 1300-2200 centipoise (cps). As the temperature increases, the viscosity decreases. As the concentration increases, the viscosity increases.

CMC has a wide variety of industrial, manufacturing and pharmacological uses. It is used as a suspending agent and viscosity modifier (thickener) to stabilize emulsions. Its high viscosity is used to make a mixture which resembles a cream or lotion.

  • In drilling muds
  • In detergents as a soil-suspending agent
  • As a chemical dispersant of oils and other carbon structures such as nanotubes
  • In resin emulsion paints, adhesives, printing inks, textile sizes
  • As protective colloid in general
  • As stabilizer in foods
  • In pharmaceuticals as a suspending agent, tablet excipient and viscosity-increasing agent
  • In the development of biostructures for drug delivery, such as biofilms, emulsions and nanoparticles

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Physical Form Powder
Quantity 3 kg
Solubility Information Soluble in aqueous solution. High viscosity CMC is soluble at up to 50mg/mL concentration but heat may be required.
Formula Weight 700kDa
Recommended Storage Store at Room Temperature (15–30°C).