missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Carboxymethylcellulose, Sodium Salt, High Viscosity

Catalog No. 217274250GM
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
217274250GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 217274250GM Supplier MilliporeSigma™ Supplier No. 217274250GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for cellulase

  • Substrate for cellulase
  • Viscosity: (2%, H2O, 25°C): 800 - 3100 mPas

Specifications

CAS 9004-32-4
Color White
Content And Storage Store at +15°C to +30°C
Quantity 250 g
Form Solid