missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carboxymethyl cellulose, sodium salt, average M.W. 90000 (DS=0.7)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Yellow |
| pH | 6.5 to 8 (1% soln.) |
| Physical Form | Granular Powder |
| Melting Point | >300.0°C |
| Quantity | 100 g |
| Infrared Spectrum | Authentic |
| Loss on Drying | 10% max. (as packed) (3 to 5 g, 105°C, 2 hrs) |
| Packaging | Glass bottle |
| Viscosity | 50-100 mPa.s 2% sol. |
| IUPAC Name | sodium;2,3,4,5,6-pentahydroxyhexanal;acetate |
| Show More |
RUO – Research Use Only