missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=1.2)

Catalog No. AC332631000
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC332631000 100 g Glass bottle
AC332630010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC332631000 Supplier Thermo Scientific Chemicals Supplier No. 332631000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
IUPAC Name sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Color White to Beige
pH 6.5 to 8 (1% soln.)
Physical Form Crystalline Powder
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 10% max. (as packed) (3 to 5 g, 105°C, 2 hrs)
Packaging Glass bottle
Merck Index 15, 1830
Viscosity 2300mPa·s, 2% sol.
Percent Purity ≥99.5%
Chemical Name or Material Carboxymethylecellulose, sodium salt, Average M.W. 250000 (DS=1.2)
Show More Show Less

RUO – Research Use Only