missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxymethylcellulose Sodium, High Viscosity, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Carboxymethylcellulose Sodium, High Viscosity, Viscosity of 1 Percent Aqueous Solution @ 25 DEG C: 1500-3000 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 12 kg |
| Loss on Drying | 10% |
| Packaging | Poly Bottle |
| Grade | USP |
| Chemical Name or Material | Carboxymethylcellulose Sodium |