man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 850568-07-9

BClOOHHOOH

CAS RN 850568-07-9

IUPAC Name: 5-chloro-2-(dihydroxyboranyl)benzoic acid
Synonyms: 5-chloro-2-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol): 200.38
Molecular Formula: C7H6BClO4
InChi Key: RENDEFHQQIYNJL-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=C(Cl)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results