accessibility menu, dialog, popup

UserName

CH3H3CCH3BrHOOSOOCH3H3COHBrCH3

CAS RN 76-59-5

IUPAC Name: 3,3-bis[3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: bromothymol blue bromthymol blue
Molecular Weight (g/mol): 624.38
Molecular Formula: C27H28Br2O5S
InChi Key: NUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=C(C)C(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(Br)=C1C
Molecular Weight (g/mol) 
  • (6)
  • (4)
  • (20)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (7)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
Grade 
  • (10)
  • (7)
  • (2)
  • (2)
  • (1)
  • (3)
Color 
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
pH 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (6)
  • (4)
  • (4)
  • (1)
  • (2)
  • (6)
  • (3)
  • (12)
  • (6)
  • (4)
  • (20)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (7)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (10)
  • (7)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)