missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Solution, Solstice

Catalog No. 6002427
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002427 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002427 Supplier Solstice Supplier No. 32822100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acid-base indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

For Use With (Application) pH Calibration
Physical Form Liquid
Quantity 100 mL
Solution Type Buffer
Flash Point 38°C (100.4°F)
UN Number NONH for all modes of transport
Beilstein 373934
Formula Weight 624.38g/mol
Grade Not Available
Concentration or Composition (by Analyte or Components) Water (7732-18-5): ≥70.00 - <90.00 %
Ethanol (64-17-5): ≥10.00 - <20.00 %
Chemical Name or Material Bromothymol Blue Solution
Show More Show Less
Precautionary Statement H226