missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Sultone Form, ACS Reagent, Reag. Ph. Eur., Solstice

Catalog No. 60046156
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046156 5 g Glass Bottle
60046155 25 g Glass Bottle
60046154 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046156 Supplier Solstice Supplier No. 327145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator, ACS Reagent, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym 3′,3″-Dibromothymolsulfonphthalein
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Packaging Glass Bottle
Suitability Complies for clarity of solution
Visual Transition Interval 5.8 to 7.4
UN Number NONH for all modes of transport
Beilstein 373934
Color Violet
Quantity 5 g
Physical Form Powder
Formula Weight 624.38g/mol
Grade ACS Reagent
Chemical Name or Material Bromothymol Blue Sultone Form, Reag. Ph. Eur.
Show More Show Less
EINECSNumber 200-971-2