missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue (0.04% in Water) [for pH Determination], TCI America™
Specifications
Specifications
| Color | Brown |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Formula Weight | 624.38 |
| Chemical Name or Material | Bromothymol Blue (0.04% in Water) [for pH Determination] |
Safety and Handling
| RTECSNumber | SJ7450000 |
| TSCA | Yes |