missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18601100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601100 500 mL Poly Bottle
18601099 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601100 Supplier Spectrum Chemical Mfg Cor Supplier No. B306500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

B-306, 34722-90-2

Spectrum™ Chemical Bromothymol Blue TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 76-59-5 , 64-17-5 , 7732-18-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.38
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
IUPAC Name 3,3-bis[3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ6-benzoxathiole-1,1-dione
SMILES CC(C)C1=CC(=C(C)C(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(Br)=C1C

Specifications

CAS Max % 0.1%, 94.90%, 5
UN Number UN1170
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL
Chemical Name or Material Bromothymol Blue TS