missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, free acid, For ACS analysis, MP Biomedicals™

Catalog No. ICN15052425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15052425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15052425 Supplier MP Biomedicals Inc Supplier No. 0215052425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym 3', 3''-Dibromothymolsulfone-phthalein
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Color Lilac
Quantity 25 g
Physical Form Powder
Formula Weight 624.4
Chemical Name or Material Bromothymol Blue, free acid, For ACS analysis