missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, 0.04% (w/v) in Methanol, Ricca Chemical

Catalog No. R1375500100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R1375500100 100 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1375500100 Supplier Ricca Chemical Company Supplier No. R1375500100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pH Indicator

Specifications

Chemical Name or Material Bromothymol Blue
Name Note 0.04% (w/v) in Methanol
CAS 76-59-5, 67-56-1
Odor Characteristic alcohol odor
Packaging Natural Poly Bottle
Molecular Formula C27H28Br2O5S
Solvent or Matrix Nonaqueous Solution
Color Orange/Red
Solubility Information Solubility in Water: Infinite
Quantity 100 mL
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Molecular Weight (g/mol) Mixture
PubChem CID 6450
ChEBI CHEBI:86155
Percent Purity 0.04% W/V
Grade Indicator
Show More Show Less