missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, TCI America™

Catalog No. B06571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B06571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B06571G Supplier TCI America Supplier No. B06571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym bromothymol blue, bromthymol blue, dibromothymolsulfophthalein, 3,3'-dibromothymolsulfonphthalein, unii-vgu4lm0h96, vgu4lm0h96, 2-bromo-4-3-3-bromo-4-hydroxy-5-isopropyl-2-methyl-phenyl-1,1-dioxo-2,1$l^ 6-benzoxathiol-3-yl-6-isopropyl-3-methyl-phenol, 2-bromo-4-3-3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl-1,1-dioxido-3h-2,1-benzoxathiol-3-yl-6-isopropyl-3-methylphenol, 4,4'-3h-2,1-benzoxathiol-3-ylidene bis 2-bromo-3-methyl-6-1-met-hylethyl phenol s,s-dioxide, bromothymol blue solution, ph 6.0-7.6
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Melting Point 202°C
Color Purple-Red
Quantity 1 g
Formula Weight 624.38
Physical Form Crystalline Powder
Chemical Name or Material Bromothymol Blue
RTECSNumber SJ7450000
TSCA Yes