missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. 01336774
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336774 10 g
01336773 1 g
01336772 25 g
01336771 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336774 Supplier Labchem Inc Supplier No. C610700010C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bromothymol Blue, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromothymol Blue, ACS Reagent (Sultone) not water soluble
CAS 76-59-5
Molecular Formula C27H28Br2O5S
Synonym Dibromothymolsulfophthalein, Bromthymol Blue
Molecular Weight (g/mol) 624.3812
Grade ACS
Packaging Glass Bottle
Specific Gravity 1
Color White
Quantity 10 g
Physical Form Solid
Show More Show Less