man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 75251-24-0

H3CCH3O

CAS RN 75251-24-0

IUPAC Name: 1-(4-ethylphenyl)propan-2-one
Synonyms: 1-(4-ethylphenyl)propan-2-one
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: DNAGWILBAXAZOH-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results