CAS RN 51527-73-2
CAS RN 51527-73-2
IUPAC Name:
2,4,6-trichlorobenzene-1-sulfonyl chloride
Synonyms:
2,4,6-trichlorobenzenesulfonyl chloride
Molecular Weight (g/mol):
279.94
Molecular Formula:
C6H2Cl4O2S
InChi Key:
WHJAQKAAIOHCGN-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C(C(Cl)=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
2,4,6-Trichlorobenzenesulfonyl Chloride 98.0+%, TCI Americaâ„¢
CAS: 51527-73-2 Molecular Formula: C6H2Cl4O2S Molecular Weight (g/mol): 279.94 MDL Number: MFCD00052973 InChI Key: WHJAQKAAIOHCGN-UHFFFAOYSA-N Synonym: 2,4,6-trichlorobenzene-1-sulfonyl chloride,2,4,6-trichlorophenylsulfonyl chloride,2,4,6-trichlorobenzenesulphonyl chloride,benzenesulfonyl chloride, 2,4,6-trichloro,2,4,6-trichlorobenzenesulfonylchloride,pubchem5175,acmc-209ku8,ksc269e6l,chloro 2,4,6-trichlorophenyl sulfone,2,4,6 trichlorobenzenesulfonyl chloride PubChem CID: 521335 IUPAC Name: 2,4,6-trichlorobenzenesulfonyl chloride SMILES: C1=C(C=C(C(=C1Cl)S(=O)(=O)Cl)Cl)Cl