man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 138008-97-6

BOCH3H3CHOOH

CAS RN 138008-97-6

IUPAC Name: [2-(propan-2-yloxy)phenyl]boronic acid
Synonyms: 2-isopropoxyphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: XDMKBIIRBDPSOE-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC=C1B(O)O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results