man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 13367-85-6

H3COHHOOO

CAS RN 13367-85-6

IUPAC Name: (5Z)-7-{2-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopent-1-en-1-yl}hept-5-enoic acid
Synonyms: prostaglandin B2
Molecular Weight (g/mol): 334.46
Molecular Formula: C20H30O4
InChi Key: PRFXRIUZNKLRHM-HKVRTXJWSA-N
SMILES: CCCCC[C@H](O)\C=C\C1=C(C\C=C/CCCC(O)=O)C(=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results