accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 13179-98-1

H3COOCH3

CAS RN 13179-98-1

IUPAC Name: 1,6-dimethoxyhexane
Synonyms: 1,6-dimethoxyhexane
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: GZHQUHIMIVEQFV-UHFFFAOYSA-N
SMILES: COCCCCCCOC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.