man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1315360-75-8

NNBrOOH

CAS RN 1315360-75-8

IUPAC Name: 3-bromoimidazo[1,2-a]pyridine-7-carboxylic acid
Synonyms: 3-bromoimidazo[1,2-a]pyridine-7-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: NBXDWHKFUACMKB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=NC=C(Br)N2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)