man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 127641-25-2

CH3HON

CAS RN 127641-25-2

IUPAC Name: 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol
Synonyms: 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol
Molecular Weight (g/mol): 205.3
Molecular Formula: C13H19NO
InChi Key: FZVHJGJBJLFWEX-UHFFFAOYNA-N
SMILES: CC(C(O)C1=CC=CC=C1)N1CCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results