CAS RN 127-65-1
CAS RN 127-65-1
IUPAC Name:
sodium chloro(4-methylbenzenesulfonyl)azanide
Synonyms:
chloramine T acti-chlore sodium tosylchloramide monochloramine T chlorozone chloralone tochlorine chloraseptine chlorasan chlorseptol
Molecular Weight (g/mol):
227.64
Molecular Formula:
C7H7ClNNaO2S
InChi Key:
VDQQXEISLMTGAB-UHFFFAOYSA-N
SMILES:
[Na+].CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl
1
–
1
of
1
results
1
Chloramine-T, 1% (w/v), Ricca Chemical
CAS: 127-65-1 Molecular Formula: C7H7ClNNaO2S Molecular Weight (g/mol): 227.638 InChI Key: VDQQXEISLMTGAB-UHFFFAOYSA-N PubChem CID: 3641960 ChEBI: CHEBI:53767 IUPAC Name: sodium;chloro-(4-methylphenyl)sulfonylazanide SMILES: CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]