man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1240257-94-6

SFFFFFH2NF

CAS RN 1240257-94-6

IUPAC Name: 2-fluoro-5-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 2-fluoro-5-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 237.16
Molecular Formula: C6H5F6NS
InChi Key: YJMWVAKSHDJWAU-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results