man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 117136-33-1

A(a)

CAS RN 117136-33-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results