man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1083168-94-8

BOOH3CCH3H3CH3CNOCH3NOO

CAS RN 1083168-94-8

IUPAC Name: 2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 280.09
Molecular Formula: C12H17BN2O5
InChi Key: XWXOPFXUTOQOSM-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=N1)B1OC(C)(C)C(C)(C)O1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results