man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 106261-48-7

CH3NNOHO

CAS RN 106261-48-7

IUPAC Name: 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
Synonyms: 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
Molecular Weight (g/mol): 234.3
Molecular Formula: C13H18N2O2
InChi Key: ZJUXJQSYXBYFFO-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=CC=C(C=C2)C(O)=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results