CAS RN 10531-80-3
CAS RN 10531-80-3
IUPAC Name:
6-methyl-1,3-benzoxazole
Synonyms:
6-methyl-1,3-benzoxazole
Molecular Weight (g/mol):
133.15
Molecular Formula:
C8H7NO
InChi Key:
SZWNDAUMBWLYOQ-UHFFFAOYSA-N
SMILES:
CC1=CC=C2N=COC2=C1
1
–
1
of
1
results
1
6-Methylbenzoxazole, 99.5+%, Thermo Scientificâ„¢
CAS: 10531-80-3 Molecular Formula: C8H7NO Molecular Weight (g/mol): 133.15 MDL Number: MFCD00216937 InChI Key: SZWNDAUMBWLYOQ-UHFFFAOYSA-N Synonym: 6-methylbenzo d oxazole,6-methylbenzoxazole,6-methyl-benzooxazole,benzoxazole, 6-methyl PubChem CID: 2724969 IUPAC Name: 6-methyl-1,3-benzoxazole SMILES: CC1=CC2=C(C=C1)N=CO2