accessibility menu, dialog, popup

UserName

CAS RN 1052236-86-8

CH2O

CAS RN 1052236-86-8

IUPAC Name: 3a-(prop-2-en-1-yl)-2H,3H,3aH,4H,5H-cyclopenta[b]furan
Synonyms: 3a-(prop-2-en-1-yl)-2H,3H,4H,5H-cyclopenta[b]furan
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: WBRYUDKSKFXYOD-UHFFFAOYNA-N
SMILES: C=CCC12CCOC1=CCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results