man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1036628-11-1

CH3ClNHF

CAS RN 1036628-11-1

IUPAC Name: 4-chloro-N-[(4-fluorophenyl)methyl]-2-methylaniline
Synonyms: 4-chloro-N-[(4-fluorophenyl)methyl]-2-methylaniline
Molecular Weight (g/mol): 249.71
Molecular Formula: C14H13ClFN
InChi Key: CYUBYUVQPNJDHU-UHFFFAOYSA-N
SMILES: CC1=C(NCC2=CC=C(F)C=C2)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results