man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 103336-06-7

H3CNOOH3CH3CCH3OOH

CAS RN 103336-06-7

IUPAC Name: 1-[(tert-butoxy)carbonyl]-2-methylpyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: YQXRKJHVAUKXRN-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1(C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results