CAS RN 1022154-80-8
CAS RN 1022154-80-8
IUPAC Name:
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-one
Synonyms:
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
Molecular Weight (g/mol):
282.09
Molecular Formula:
C14H17BF2O3
InChi Key:
NDJMQXWRZUTNSA-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(F)C=C(B2OC(C)(C)C(C)(C)O2)C(F)=C1
1
–
1
of
1
results
1
4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1022154-80-8 Molecular Formula: C14H17BF2O3 Molecular Weight (g/mol): 282.094 MDL Number: MFCD22419259 InChI Key: NDJMQXWRZUTNSA-UHFFFAOYSA-N Synonym: 4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester,1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone PubChem CID: 68668352 IUPAC Name: 1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F