accessibility menu, dialog, popup

UserName

Check Out the New Spotlight Offers and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1005-67-0

H3CNO

CAS RN 1005-67-0

IUPAC Name: 4-butylmorpholine
Synonyms: morpholine, 4-butyl-
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: LMRKVKPRHROQRR-UHFFFAOYSA-N
SMILES: CCCCN1CCOCC1
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.