Learn More
MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns
PEEK coated HPLC column
Supplier:  MilliporeSigma™ 1504510001
 Afficher plus de versions de ce produit
Numéro de catalogue. M1504500001
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more
Spécifications
Spécifications
4.6 mm
5 μm
200 Å
Metal-free PEEK
50 mm
200 Å
1/Pk.
Column
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS