missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns

Catalog No. p-4752744
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
Particle Size:
3.5 μm
5 μm
Porosity:
100 Å
200 Å
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size Porosity
M1504440001 150 mm 4.6 mm 3.5 μm 100 Å
M1504420001 150 mm 2.1 mm 3.5 μm 100 Å
M1504500001 50 mm 4.6 mm 5 μm 200 Å
M1504540001 150 mm 2.1 mm 5 μm 200 Å
M1504520001 100 mm 2.1 mm 5 μm 200 Å
M1504480001 150 mm 2.1 mm 3.5 μm 200 Å
M1504530001 100 mm 4.6 mm 5 μm 200 Å
M1504400001 50 mm 2.1 mm 3.5 μm 100 Å
M1504550001 150 mm 4.6 mm 5 μm 200 Å
M1504490001 150 mm 4.6 mm 3.5 μm 200 Å
M1504430001 250 mm 2.1 mm 3.5 μm 100 Å
M1504570001 250 mm 2.1 mm 5 μm 200 Å
M1504460001 50 mm 4.6 mm 3.5 μm 200 Å
M1504470001 100 mm 2.1 mm 3.5 μm 200 Å
M1504450001 50 mm 2.1 mm 3.5 μm 200 Å
M1504580001 250 mm 4.6 mm 5 μm 200 Å
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. M1504440001 Supplier MilliporeSigma™ Supplier No. 1.50444.0001
Only null left

PEEK coated HPLC column

SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Particle Size 3.5 μm
Pore Size 100 Å
Porosity 100 Å
Quantity 1/Pk.
Material Metal-free PEEK
Type Column