Learn More
MilliporeSigma™ Express™ PLUS Membrane Filters
Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.
Supplier: MilliporeSigma™ HPWP02500
Description
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
Features and Benefit:- Ultra fast filtration and high throughput shorten process time
- Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
- Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Spécifications
Not Autoclavable | |
Reusable | |
Disc | |
PES | |
Non-sterile | |
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices. | |
Filter Membranes | |
1 in. |
White | |
Screen Filter | |
PES | |
0.45 μm | |
23 (at 690 Mbar (10 psi)) | |
Hydrophilic | |
Hydrophilic | |
25 mm |