UserName
Analytical Testing Methods
Analytical Testing Methods
High-Quality Analytical Testing Products from Top Brands
Analytical testing methods help Iabs and researchers identify and monitor acidity and ion concentration, water content, and many other properties of a range of sample types.
Karl Fischer titration, pH, and ASTM methods are frequently used analytical techniques used during water testing and environmental analysis, food and beverage testing, pharmaceutical production, production of cosmetics, petroleum, building materials, and more, as well as clinical research.
Accurate, consistent outcomes are mandatory, and efficiency is just as important. The analytical testing chemicals and lab products you choose help deliver the outcomes you need, and boost confidence that your samples won't be wasted.
Save on Select Electrochemistry Products
Find Your Testing Method
Check out the methods below to explore products and resources to help meet your analytical testing needs.
pH & Electrochemistry
pH and electrochemistry meters and related products are used to assess acidity and alkalinity, conductivity, specific ion concentrations, and dissolved oxygen in water, wastewater, and other solutions.
Electrodes must be carefully stored and maintained for optimal performance. It’s important to select the right storage bottles and electrode filling, cleaning, rinsing, and storage accessories.
Electrochemistry meters also require regular calibration and maintenance. Standard and certified pH buffers as well as conductivity and specific ion standards are available. Other accessories include electrode supports, bulb guards, cables, connectors, and adapters.
Top Categories
Resources
Filename
thermo-scientific-orion-benchtop-portable-meters-brochure.pdf
Size
3 MB
Format
application/pdf
Filename
labchem-products-regulated-testing-datasheet.pdf
Size
2 MB
Format
application/pdf
Filename
labchem-environmental-testing-solutions-datasheet.pdf
Size
2 MB
Format
application/pdf
Filename
electrochemistry-workflow-products-data-sheet-us-en-20-487-2293.pdf
Size
668 KB
Format
application/pdf
Filename
labchem-analytical-testing-reagent-data-sheet.pdf
Size
695 KB
Format
application/pdf
Karl Fischer Titration
Karl Fischer titration is used with a range of sample types to determine trace amounts of water in samples. Labs and researchers use two types of Karl Fischer titration based on a sample's properties.
Coulometric KF titration is ideal for liquid and gaseous samples with low water content (up to 2%). Water content is measured by electrical current, and the instrument's cell is made from glass with grounded joint connections that ensure a tight seal against air moisture.
Volumetric KF titration is used with solid, liquid and gaseous samples with high water content (over 2%). Water content is measured by burette instead of electrical current, and the instrument's cell construction differs from a coulometric titrator.
Top Categories
Resources
Filename
fisher-chemical-aqualine-reagents-brochure.pdf
Size
1 MB
Format
application/pdf
Filename
ricca-chemical-hydrospec-reagents-brochure.pdf
Size
872 KB
Format
application/pdf
Filename
honeywell-hydranal-karl-fischer-titration-us-en.pdf
Size
433 KB
Format
application/pdf
Filename
honeywell-research-chemicals-brochure.pdf
Size
1 MB
Format
application/pdf
ASTM Methods Products
ASTM test methods are definitive procedures that produce a result. They usually include a detailed description of a procedure for determining a property or constituent of a material, an assembly of materials, or a product. These details include the apparatus, test specimen, procedure, and calculations needed for satisfactory results.
ASTM test methods represent a consensus about the best currently available procedure and are supported by experience and data from cooperative tests.
The ASTM publishes six types of standards: test method, specification, classification, practice, guide, and terminology.
Learn More About ASTM Testing Standards
Resources
Filename
four-w's-of-astm-datasheet.pdf
Size
285 KB
Format
application/pdf
Filename
fisher-chemical-new-solvent-grade-targeted-for-trace-analysis-uhplc-ms.pdf
Size
551 KB
Format
application/pdf
Filename
fisher-chemical-tracemetal-grade-acids-and-base-brochure.pdf
Size
1 MB
Format
application/pdf
Filename
labchem-solutions-reagents-method-chemistry-flier.pdf
Size
165 KB
Format
application/pdf
Filename
thermo-scientific-orion-benchtop-portable-meters-brochure1.pdf
Size
3 MB
Format
application/pdf
Learn More
Safety and Lab Essentials
Your people and the work they do are your biggest investments. Help keep them safe and productive with innovative safety products from the top brands. From face and hand protection to spill containment and cleanup, the Fisher Scientific channel has the products you need to prepare, prevent, protect, and respond.
Whether you need glass, plastic, or both, you can find a range of suitable lab glassware and plasticware varieties to help conduct your experiments. Choose from multiple products, materials, sizes, packaging, and more.
Top Categories
Resources
Filename
fisher-scientific-chemical-splash-exposure-brochure.pdf
Size
4 MB
Format
application/pdf
Learn More