CAS RN 9005-65-6
Tween™ 80, Fisher BioReagents
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N Synonym: Polysorbate 80,Polyoxyethylene-20-sorbitan Monooleate IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Tween 80, MP Biomedicals
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Tween 80 (=Polyoxyethylene Sorbitan Monooleate), TCI America™
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N Synonym: Polyoxyethylene Sorbitan Monooleate IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 80, Multi-Compendial, N.F., J.T. Baker™
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polyoxyethylene (20) Sorbitan Monooleate, BAKER™, J.T. Baker™
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 80, Vegetable, Multi-Compendial, N.F., J.T. Baker™
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 80
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N Synonym: Polyoxyethylene sorbitan monooleate IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MilliporeSigma™ Tween™ 80 (Polyoxyethylene (20) Monooleate), OmniPur™, Calbiochem™,
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N Synonym: Polyoxyethylene Sorbitan Monooleate IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Triethanolamine, 97 to 98%, MP Biomedicals™
CAS: 9005-65-6
Tween 80 (=Polyoxyethylene Sorbitan Monooleate) [for Biochemical Research], TCI America™
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N Synonym: Polyoxyethylene Sorbitan Monooleate IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 80, NF, Spectrum™ Chemical
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 80, NF, Spectrum™ Chemical
CAS: 9005-65-6 Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 Molecular Weight (g/mol): 522.68 MDL Number: MFCD00165986 InChI Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Polysorbate 80, FCC, 65-69.5%, Spectrum™ Chemical
CAS: 9005-65-6 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 Molecular Weight (g/mol): 604.82 MDL Number: MFCD00082107 InChI Key: HDTIFOGXOGLRCB-KTKRTIGZNA-N IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
- 1
- 2