missing translation for 'onlineSavingsMsg'
Learn More

Tween 80 (=Polyoxyethylene Sorbitan Monooleate), TCI America™

Catalog No. T0546500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0546500G 500 g
T054625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0546500G Supplier TCI America Supplier No. T0546500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
Synonym Polyoxyethylene Sorbitan Monooleate
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Clear Liquid at 20°C
Melting Point -25°C
pH 5 to 7 (5% H2O soln.)
Quantity 500 g
Chemical Name or Material Tween 80 (=Polyoxyethylene Sorbitan Monooleate)
EINECSNumber (8)-0055
RTECSNumber WG2932500
TSCA Yes