missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 97 to 98%, MP Biomedicals™

Catalog No. MP215216390
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215216390 500 mL
ICN15216380 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215216390 Supplier MP Biomedicals Inc Supplier No. 0215216390

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Triethanolamine ≥97%, Liquid

  • It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks.
  • It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating.
  • It is used in hair care products like hair dyes and wave sets.
  • Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions.

Chemical Identifiers

CAS 9005-65-6

Specifications

Melting Point 17.9°C to 21°C (literature)
Density 1.124g/cm3 at 20°C(Lit.)
pH 9 to 11.5 (1% aq soln) useful pH range 7.3 to 8
Boiling Point 190°C to 193°C (5 mmHg) (Literature)
Flash Point ca. 190°C (closed cup)(Lit.)
Molecular Formula C6H15NO3
Refractive Index n20/D 1.485(lit.)
MDL Number MFCD00002855
Quantity 500 mL
Beilstein 1699263
Synonym Daltogen, Nitrilo-2,2,2-Triethanol, Sterolamide, Trihydroxytriethylamine
Solubility Information Soluble in water (30% w/v - clear, colorless solution); miscible in methanol, acetone, benzene, ether.
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
SMILES OCCN(CCO)CCO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Molecular Weight (g/mol) 149.19
Vapor Density 5.14 (air = 1)(Lit.)
Vapor Pressure 0.000000477 hPa at 25°C(Lit.)
Physical Form Liquid
Grade ≥97%
Show More Show Less
RTECSNumber KL9275000
storageNote1 Room Temperature, desiccate, protect from light, store under nitrogen