missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80, Vegetable, Multi-Compendial, N.F., J.T. Baker™

Catalog No. 02003663
Change view
Click to view available options
Quantity:
200 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02003663 4 L
02003655 200 mL
02003660 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02003663 Supplier Avantor Supplier No. 450003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 4 L
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.08kg
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 80, Vegetable