missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tween™ 80 (Polyoxyethylene (20) Monooleate), OmniPur™, Calbiochem™,

Catalog No. 94901L
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
94901L 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 94901L Supplier MilliporeSigma™ Supplier No. 94901L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for solubilization of membrane proteins, enzymology, analytical and diagnostic work, as well as chromatography and electrophoresis

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
Synonym Polyoxyethylene Sorbitan Monooleate
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Viscous liquid
Quantity 1 L
Packaging Plastic Bottle
Grade Molecular Biology