missing translation for 'onlineSavingsMsg'
Learn More

Tween 80, MP Biomedicals

Catalog No. ICN10317080
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10317080 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10317080 Supplier MP Biomedicals Inc Supplier No. 0210317080
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

9005-65-6 (Polyoxyethylenesorbitan monooleate) Syrup Purity: ∼75% oleic acid

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 100 mL
Chemical Name or Material Tween 80