accessibility menu, dialog, popup

UserName

KONO

CAS RN 7758-09-0

IUPAC Name: potassium nitrite
Synonyms: potassium nitrite
Molecular Weight (g/mol): 85.1
Molecular Formula: KNO2
InChi Key: BXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES: [K+].[O-]N=O
Grade 
  • (4)
  • (6)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Physical Form 
  • (5)
  • (8)
  • (4)
Percent Purity 
  • (3)
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (8)
  • (2)
  • (1)
  • (2)
  • (7)
  • (8)
  • (11)
  • (4)
  • (6)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)
  • (8)
  • (4)
  • (3)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)