missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Nitrogen Standard, 1000 ppm N (3285 ppm NO2-), Ricca Chemical

Catalog No. 54614
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
54614 120 mL Natural Poly Bottle
546116 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54614 Supplier Ricca Chemical Company Supplier No. 54614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-09-0 , 7732-18-5
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 0.62
CAS Max % 100.0, 0.65
Analyte or Component Names KNO2- in water
Equivalence 1 mL = 1 mg N as NO2
pH Approximately 7
Boiling Point 100°C
Element Multi-element
Solvent or Matrix Water
Quantity 120 mL
Grade ASTM
Concentration or Composition (by Analyte or Components) 1000ppm N, 3285ppm NO2-
Chemical Name or Material Nitrogen Standard (as Nitrite)
Show More Show Less
ShelfLife 6 month shelf life refrigerated.