missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrite, 97%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 387.0°C |
| Color | White to Yellow |
| Physical Form | Crystals |
| Quantity | 250 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 97% |
| Linear Formula | KNO2 |
| Fieser | 10,329 |
| Merck Index | 15, 7770 |
| Solubility Information | Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Toxic if swallowed. Very toxic to aquatic life. May intensify fire; oxidizer. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 231-832-4 |
RUO – Research Use Only