missing translation for 'onlineSavingsMsg'
Learn More

Nitrite (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 3.28mg NO2- ), LabChem™

Catalog No. LC179207
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC179207 125 mL Poly Bottle
LC179201 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC179207 Supplier LabChem Supplier No. LC179207

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-09-0 , 7732-18-5
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Packaging Poly Bottle
CAS Max % 99.39, 0.61
Identification Passes Test
Density 1g/mL
Specific Gravity 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm
Linear Formula KNO2
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 85.1
Grade Certified
Assay as N
Chemical Name or Material Nitrite (as Nitrogen) Standard, 1000 ppm, (1mL = 1mg N, 1mL = 3.28mg NO2-)
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Refrigerate