missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Nitrite, Crystal, Technical, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Crystals or Needles |
| CAS Max % | 1 |
| Quantity | 12 kg |
| Packaging | Amber Glass Bottle |
| Molecular Weight (g/mol) | 85.10 |
| Grade | Technical |
| Chemical Name or Material | Potassium Nitrite, Crystal |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.